missing translation for 'onlineSavingsMsg'
Learn More

Oxetan-3-ol, 95%

Product Code. p-4228859 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10799400

Brand: Thermo Scientific Acros 435840010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7748-36-9 | C3H6O2 | 74.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7748-36-9
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
MDL Number MFCD09056790
InChI Key QMLWSAXEQSBAAQ-UHFFFAOYSA-N
Synonym 3-hydroxyoxetane, 3-oxetanol, 1,3-epoxy-2-propanol, oxetane-3-ol, 3-hydroxy-oxetane, 3-hydroxy oxetane, pubchem17367, ksc380k3t, 1,3-epoxypropan-2-ol
PubChem CID 9942117
IUPAC Name oxetan-3-ol
SMILES OC1COC1

Specifications

Color Colorless
Density 1.1250g/mL
Boiling Point 153°C
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.125
Formula Weight 74.08
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Oxetan-3-ol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Oxetan-3-ol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.