missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXER1 Polyclonal Antibody

OXER1 Polyclonal Antibody
Brand: Invitrogen PA575410
This item is not returnable.
View return policy
Description
OXER1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| OXER1 | |
| Polyclonal | |
| Unconjugated | |
| OXER1 | |
| 5-oxo-ETE acid G-protein-coupled receptor 1; 5-oxo-ETE G-protein coupled receptor; GPCR; GPR170; G-protein coupled receptor 170; G-protein coupled receptor R527; G-protein coupled receptor TG1019; HEOAD54; OXE receptor; OXER1; oxoeicosanoid (OXE) receptor 1; oxoeicosanoid receptor 1; TG1019 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 165140 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TDS5 | |
| OXER1 | |
| Synthetic peptide corresponding to amino acids 220-270 of Human OXER1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction