missing translation for 'onlineSavingsMsg'
Learn More

Oxeladin N-Oxide, Mikromol™

Product Code. 30050043
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050043 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050043 Supplier Mikromol Supplier No. MM0547.01
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Oxeladin N-Oxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxeladin N-Oxide
CAS 1812854-79-7
Analyte or Component Names Oxeladin N-Oxide
Shipping Condition Ice pack (-20°C)
Molecular Formula C20 H33 N O4
Certifications/Compliance ISO 9001
Synonym Oxeladin N-Oxide
InChI Formula InChI=1S/C20H33NO4/c1-5-20(6-2,18-12-10-9-11-13-18)19(22)25-17-16-24-15-14-21(23,7-3)8-4/h9-13H,5-8,14-17H2,1-4H3
SMILES CCC(CC)(C(=O)OCCOCC[N+]([O-])(CC)CC)c1ccccc1
IUPAC Name N,N-diethyl-2-[2-(2-ethyl-2-phenylbutanoyl)oxyethoxy]ethanamine oxide
Molecular Weight (g/mol) 351.48
Quantity 10 mg
Formula Weight 351.241 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.