missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OXCT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116604
This item is not returnable.
View return policy
Description
Positive Samples: Mouse testis, Rat testis Immunogen sequence: QKTRVVVTMQ HCTKDNTPKI MEKCTMPLTG KRCVDRIITE KAVFDVHRKK ELTLRELWEG LTVDDIKKST GCAFAVSPNL R.
Key enzyme for ketone body catabolism. Transfers the CoA moiety from succinate to acetoacetate. Formation of the enzyme-CoA intermediate proceeds via an unstable anhydride species formed between the carboxylate groups of the enzyme and substrate.
Specifications
| OXCT2 | |
| Polyclonal | |
| Unconjugated | |
| OXCT2 | |
| 3-oxoacid CoA transferase 2; 3-oxoacid CoA transferase 2A; 3-oxoacid CoA-transferase 2; 3-oxoacid CoA-transferase 2A; 3-oxoacid-CoA transferase 2A; FKSG25; haploid germ cell specific succinyl CoA transferase; Oxct; Oxct2; Oxct2a; Scot; SCOTT; SCOT-t; SCOT-t1; succinyl-CoA:3-ketoacid coenzyme A transferase 2, mitochondrial; Succinyl-CoA:3-ketoacid coenzyme A transferase 2A, mitochondrial; succinyl-CoA:3-ketoacid-coenzyme A transferase 2, mitochondrial; succinyl-CoA:3-ketoacid-coenzyme A transferase 2A, mitochondrial; succinyl-CoA:3-oxoacid CoA-transferase; testis-specific succinyl CoA:3-oxoacid CoA-transferase; testis-specific succinyl CoA:3-oxoacid CoA-transferase 1; testis-specific succinyl-CoA:3-oxoacid CoA-transferase; Testis-specific succinyl-CoA:3-oxoacid CoA-transferase 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 366463, 64059, 64064 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.65 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q5XIJ9, Q9BYC2, Q9JJN4 | |
| OXCT2, Oxct2a | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 430-510 of human OXCT2 (NP_0714031). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction