Learn More
OXCT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543183
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: GMYANLGIGI PLLASNFISP NITVHLQSEN GVLGLGPYPR QHEADADLIN Sequence homology: Cow: 100%; Dog: 92%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 100%; Zebrafish: 92%
OXCT1 Polyclonal antibody specifically detects OXCT1 in Human, Rat samples. It is validated for Western Blot.Specifications
OXCT1 | |
Polyclonal | |
Unconjugated | |
OXCT1 | |
2610008O03Rik; 3-oxoacid CoA transferase 1; 3-oxoacid CoA-transferase 1; 3-oxoacid-CoA transferase 1; EMBL:AAI66478.1}; OXCT; Oxct1; oxct1 {ECO:0000312; Oxct2a; SCOT; SCOT-s; somatic-type succinyl CoA:3-oxoacid CoA-transferase; somatic-type succinyl-CoA:3-oxoacid CoA-transferase; somatic-type succinyl-CoA:3-oxoacid-CoA-transferase; succinyl CoA:3-oxoacid CoA transferase; succinyl-CoA:3-ketoacid CoA transferase; succinyl-CoA:3-ketoacid coenzyme A transferase 1, mitochondrial; succinyl-CoA:3-ketoacid-CoA transferase; succinyl-CoA:3-ketoacid-coenzyme A transferase 1, mitochondrial; Succinyl-CoA:3-ketoacid-coenzyme A transferase 1, mitochondrial-like protein; Unknown (protein for MGC:137519) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
5019, 690163 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
B2GV06, P55809 | |
OXCT1 | |
Synthetic peptide directed towards the middle region of human OXCT1 | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |