Learn More
OXCT1 Mouse, anti-Human, Unconjugated, Clone: 1G1B9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67836-1-IG-150UL
Description
OXCT1 (also know as SCOT), encoded by nuclear gene, is a mitochondrial CoA transferase required for ketone body degradation. It catalyzes the transfer of CoA from succinyl-CoA to acetoacetate, generating acetoacetyl-CoA. OXCT1 is expressed in brain, heart, and skeletal muscle, but not in liver. This antibody specifically recognizes endogenous OXCT1. (21209089)
Specifications
| OXCT1 | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse, Rabbit, Pig |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1G1B9 | |
| Unconjugated | |
| 150 μL | |
| 5019 | |
| Liquid |