missing translation for 'onlineSavingsMsg'
Learn More

Oxcarbazepine EP Impurity C (Oxcarbazepine USP Related Compound E), Mikromol™

Product Code. 30459943
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459943 4 x 25 mg -
30052321 25 mg 25mg
30050641 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30459943 Supplier Mikromol Supplier No. MM0076.1400254AMP
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Oxcarbazepine EP Impurity C (Oxcarbazepine USP Related Compound E), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 10-Oxo-10,11-Dihydro-5H-dibenz[b,f]azepine
CAS 21737-58-6
Analyte or Component Names 10-Oxo-10,11-Dihydro-5H-dibenz[b,f]azepine
Shipping Condition Room Temperature
Molecular Formula C14 H11 N O
Certifications/Compliance ISO 17025
Synonym 10-Ketoiminodibenzyl (10-Oxo-10,11-dihydro-5H-dibenz[b,f]azepine), Oxcarbazepine Imp. C (EP), Oxcarbazepine USP Related Compound E, Oxcarbazepine USP RC E, 5,11-Dihydro-10H-dibenz[b,f]azepin-10-one, Oxcarbazepine Related Compound E, Oxcarbazepine Impurity C
InChI Formula InChI=1S/C14H11NO/c16-14-9-10-5-1-3-7-12(10)15-13-8-4-2-6-11(13)14/h1-8,15H,9H2
Solution Type Neat
SMILES O=C1Cc2ccccc2Nc3ccccc13
IUPAC Name 6,11-dihydrobenzo[b][1]benzazepin-5-one
Molecular Weight (g/mol) 209.24
Quantity 4 x 25 mg
Formula Weight 209.0841 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.