missing translation for 'onlineSavingsMsg'
Learn More

Oxazole, 98+%

Product Code. 11394955
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394955 5 g 5g
11384955 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394955 Supplier Thermo Scientific Alfa Aesar Supplier No. L18341.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 288-42-6 | C3H3NO | 69.063 g/mol

Oxazole and its derivatives are actively involved as building blocks for bio chemicals and pharmaceuticals like flopristin and darglitazone. It is also involved in industrial applications such as in dyes, fluorescent brightening agents, textile auxiliaries and plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxazole and its derivatives are actively involved as building blocks for bio chemicals and pharmaceuticals like flopristin and darglitazone. It is also involved in industrial applications such as in dyes, fluorescent brightening agents, textile auxiliaries and plastics.

Solubility
Miscible with alcohol and ether. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-42-6
Molecular Formula C3H3NO
Molecular Weight (g/mol) 69.063
MDL Number MFCD00009751
InChI Key ZCQWOFVYLHDMMC-UHFFFAOYSA-N
Synonym oxazole, unii-fjz20i1lps, fjz20i1lps, oxazol, 1,3-oxazol, pubchem8626, oxazole 1g
PubChem CID 9255
ChEBI CHEBI:35597
IUPAC Name 1,3-oxazole
SMILES C1=COC=N1

Specifications

Melting Point -86°C
Density 1.05
Boiling Point 69°C to 70°C
Flash Point 19°C (66°F)
Refractive Index 1.427
Quantity 5 g
UN Number UN2924
Beilstein 103851
Solubility Information Miscible with alcohol and ether. Slightly miscible with water.
Formula Weight 69.07
Percent Purity ≥98%
Chemical Name or Material Oxazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Oxazole
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Oxazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.