missing translation for 'onlineSavingsMsg'
Learn More

Oxaprozin, MedChemExpress

Product Code. 16478628
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478628 10 mg 10mg
16458466 100 mg 100mg
16444497 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478628 Supplier Medchem Express Supplier No. HYB0808
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Oxaprozin is an orally active and potent COX inhibitor, with IC50 values of 2.2 μM for human platelet COX-1 and and 36 μM for IL-1-stimulated human synovial cell COX-2, respectively. Oxaprozin also inhibits the activation of NF-κB. Oxaprozin induces cell apoptosis. Oxaprozin shows anti-inflammatory activity. Oxaprozin-mediated inhibition of the Akt/IKK/NF-κB pathway contributes to its anti-inflammatory properties.

Oxaprozin is an orally active and potent COX inhibitor, with IC50 values of 2.2 μM for human platelet COX-1 and and 36 μM for IL-1-stimulated human synovial cell COX-2, respectively. Oxaprozin also inhibits the activation of NF-κB. Oxaprozin induces cell apoptosis. Oxaprozin shows anti-inflammatory activity. Oxaprozin-mediated inhibition of the Akt/IKK/NF-κB pathway contributes to its anti-inflammatory properties.

Specifications

Chemical Name or Material Oxaprozin
Physical Form Solid
CAS 21256-18-8
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C18H15NO3
For Use With (Application) COVID-19-immunoregulation
Synonym Oxaprozinum Wy21743
Solubility Information DMSO : ≥ 100 mg/mL (340.92 mM)
SMILES O=C(O)CCC1=NC(C2=CC=CC=C2)=C(C3=CC=CC=C3)O1
Molecular Weight (g/mol) 293.32
Formula Weight 293.32
Percent Purity 99.76%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.