missing translation for 'onlineSavingsMsg'
Learn More

Oxantel Pamoate, Mikromol™

Product Code. 17992826
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17992826

Brand: Mikromol MM0893.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Oxantel Pamoate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxantel Pamoate
CAS 68813-55-8
Analyte or Component Names Oxantel Pamoate
Shipping Condition Room Temperature
Molecular Formula C23 H16 O6 . C13 H16 N2 O
Certifications/Compliance ISO 17034
Synonym 2-Naphthalenecarboxylic acid, 4,4'-methylenebis[3-hydroxy-, compd. with 3-[(1E)-2-(1,4,5,6-tetrahydro-1-methyl-2-pyrimidinyl)ethenyl]phenol (1:1), 2-Naphthalenecarboxylic acid, 4,4'-methylenebis[3-hydroxy-, compd. with (E)-3-[2-(1,4,5,6-tetrahydro-1-methyl-2-pyrimidinyl)ethenyl]phenol (1:1), Phenol, 3-[(1E)-2-(1,4,5,6-tetrahydro-1-methyl-2-pyrimidinyl)ethenyl]-, 4,4'-methylenebis[3-hydroxy-2-naphthalenecarboxylate] (1:1) (salt) (9CI), Phenol, 3-[2-(1,4,5,6-tetrahydro-1-methyl-2-pyrimidinyl)ethenyl]-, (E)-, 4,4'-methylenebis[3-hydroxy-2-naphthalenecarboxylate] (1:1) (salt), CP 14445-16, Oxantel ebonate, Oxantel pamoate, Telopar
InChI Formula InChI=1S/C23H16O6.C13H16N2O/c24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;1-15-9-3-8-14-13(15)7-6-11-4-2-5-12(16)10-11/h1-10,24-25H,11H2,(H,26,27)(H,28,29);2,4-7,10,16H,3,8-9H2,1H3/b;7-6+
SMILES CN1CCCN=C1\C=C\c2cccc(O)c2.OC(=O)c3cc4ccccc4c(Cc5c(O)c(cc6ccccc56)C(=O)O)c3O
IUPAC Name 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid;3-[(E)-2-(1-methyl-5,6-dihydro-4H-pyrimidin-2-yl)ethenyl]phenol
Molecular Weight (g/mol) 604.65
Quantity 250 mg
Formula Weight 604.221 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.