missing translation for 'onlineSavingsMsg'
Learn More

Oxamide, 98%, Thermo Scientific Chemicals

Catalog Number 10607102
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
471-46-5
C2H4N2O2
88.06
MFCD00008007
YIKSCQDJHCMVMK-UHFFFAOYSA-N
ethanediamide, oxalamide, oxamimidic acid, oxalic acid diamide, diaminoglyoxal, oxamid, amid kyseliny stavelove, formimidic acid, 1-carbamoyl, oxamid czech, unii-sbe4m0223e
10113
CHEBI:48248
oxamide
C(=O)(C(=O)N)N
This item is not returnable. View return policy
471-46-5
C2H4N2O2
88.06
MFCD00008007
YIKSCQDJHCMVMK-UHFFFAOYSA-N
ethanediamide, oxalamide, oxamimidic acid, oxalic acid diamide, diaminoglyoxal, oxamid, amid kyseliny stavelove, formimidic acid, 1-carbamoyl, oxamid czech, unii-sbe4m0223e
10113
CHEBI:48248
oxamide
C(=O)(C(=O)N)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 491°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula H2NCOCONH2
Quantity 100 g
Beilstein 02, 545
Merck Index 15, 7017
Solubility Information Solubility in water: insoluble in water. Other solubilities: sparingly soluble in hot water,alcohol and ether
Formula Weight 88.06
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Oxamide
Show More Show Less
compliance-icons
Product Identifier
  • Oxamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'