missing translation for 'onlineSavingsMsg'
Learn More

Oxamic hydrazide, 99%

Product Code. 10755241 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10755241 100 g Plastic bottle 100g
10696902 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10755241 Supplier Thermo Scientific Acros Supplier No. 129641000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 515-96-8 | C2H5N3O2 | 103.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 515-96-8
Molecular Formula C2H5N3O2
Molecular Weight (g/mol) 103.08
MDL Number MFCD00008004
InChI Key MOKRDWKSHLLYKM-UHFFFAOYSA-N
Synonym oxamic hydrazide, semioxamazide, semioxamazid, aminooxamide, oxamic acid hydrazide, oxamichydrazide, n-aminooxamide, 1-hydrazinecarbonyl formamide, oxamic acid, hydrazide, unii-bet2ubd6nv
PubChem CID 68200
IUPAC Name 2-hydrazinyl-2-oxoacetamide
SMILES C(=O)(C(=O)NN)N

Specifications

Melting Point 216.0°C to 218.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula H2NCOCONHNH2
Quantity 100 g
Beilstein 02, 559
Merck Index 15, 8584
Formula Weight 103.08
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Oxamic hydrazide, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.