missing translation for 'onlineSavingsMsg'
Learn More

Oxamic Acid, TRC

Product Code. 30419462
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30419462 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419462 Supplier Toronto Research Chemicals Supplier No. TRCO8451601G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxamic Acid
CAS 471-47-6
Molecular Formula C2H3NO3
Quantity 1 g
Synonym Aminooxo-acetic Acid, 2-Amino-2-oxoacetic Acid, Aminooxoacetic Acid, (Aminocarbonyl)-formic Acid, 2-Oxo-glycine, NSC 47001, NSC 49416, NSC 53102, Oxalamic Acid, Oxalic Acid monoamide, Oxamidic Acid
InChI Formula InChI=1S/C2H3NO3/c3-1(4)2(5)6/h(H2,3,4)(H,5,6)
SMILES C(=O)(C(=O)O)N
Molecular Weight (g/mol) 89.05
Formula Weight 89.0113
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.