missing translation for 'onlineSavingsMsg'
Learn More

Oxamic acid, 98%

Product Code. 11474227
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474227 100 g 100g
11454227 5 g 5g
11464227 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474227 Supplier Thermo Scientific Alfa Aesar Supplier No. B20344.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 471-47-6 | C2H3NO3 | 89.05 g/mol

Oxamic acid could also inhibit lactic dehydrogenase in the intact Ehrlich ascites tumor cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxamic acid could also inhibit lactic dehydrogenase in the intact Ehrlich ascites tumor cell.

Solubility
Soluble in water 108 mg/mL.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-47-6
Molecular Formula C2H3NO3
Molecular Weight (g/mol) 89.05
MDL Number MFCD00008006
InChI Key SOWBFZRMHSNYGE-UHFFFAOYSA-N
Synonym 2-amino-2-oxoacetic acid, oxamate, acetic acid, aminooxo, oxalamic acid, oxamidic acid, glycine, 2-oxo, oxalic acid monoamide, amino oxo acetic acid, aminooxoacetic acid, oxamate repellent
PubChem CID 974
ChEBI CHEBI:18058
IUPAC Name oxamic acid
SMILES C(=O)(C(=O)O)N

Specifications

Melting Point ∼205°C (decomposition)
Quantity 100 g
Beilstein 1743294
Merck Index 14,6917
Solubility Information Soluble in water 108mg/mL.
Formula Weight 89.05
Percent Purity 98%
Chemical Name or Material Oxamic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.