missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | Oxalyl Chloride |
| CAS | 79-37-8 |
| Molecular Formula | C2Cl2O2 |
| Quantity | 250 g |
| Synonym | Ethanedioyl Dichloride, Oxalic Acid Chloride, Oxalic Acid Dichloride, Oxalic Dichloride, Oxaloyl Chloride, Oxaloyl Dichloride, Oxalyl Dichloride |
| InChI Formula | InChI=1S/C2Cl2O2/c3-1(5)2(4)6 |
| SMILES | ClC(=O)C(=O)Cl |
| IUPAC Name | oxalyl dichloride |
| Molecular Weight (g/mol) | 126.93 |
| Formula Weight | 125.9275 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Vous avez repéré une opportunité d'amélioration ?