missing translation for 'onlineSavingsMsg'
Learn More

Oxalyl Chloride, TRC

Product Code. 30421819
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30421819 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30421819 Supplier Toronto Research Chemicals Supplier No. TRCO845020100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxalyl Chloride
CAS 79-37-8
Molecular Formula C2Cl2O2
Quantity 100 g
Synonym Ethanedioyl Dichloride, Oxalic Acid Chloride, Oxalic Acid Dichloride, Oxalic Dichloride, Oxaloyl Chloride, Oxaloyl Dichloride, Oxalyl Dichloride
InChI Formula InChI=1S/C2Cl2O2/c3-1(5)2(4)6
SMILES ClC(=O)C(=O)Cl
IUPAC Name oxalyl dichloride
Molecular Weight (g/mol) 126.93
Formula Weight 125.9275
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.