missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid, TRC

Product Code. 30004435
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
Unit Size:
250g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004435 250 g 250g
30006979 5 g 5g
30004864 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004435 Supplier Toronto Research Chemicals Supplier No. TRCO845120250G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-62-7
Molecular Formula C2 H2 O4
Molecular Weight (g/mol) 90.03
Synonym Oxalic Acid, Ethanedioic acid, Oxalic acid, Aktisal, Aquisal, DeerClean, NSC 132055, NSC 151956, NSC 62774, NSC 76990, Ultraplast Activate S 52
IUPAC Name oxalic acid
SMILES OC(=O)C(=O)O

Specifications

InChI Formula InChI=1S/C2H2O4/c3-1(4)2(5)6/h(H,3,4)(H,5,6)
Formula Weight 89.9953
Percent Purity >95%
Quantity 250 g
Chemical Name or Material Oxalic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.