missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid Potassium Salt Dihydrate, TRC

Product Code. 30428503
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428503 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428503 Supplier Toronto Research Chemicals Supplier No. TRCO8451255G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxalic Acid Potassium Salt Dihydrate
CAS 6100-20-5
Molecular Formula C2 H2 O4 . C2 H O4 . K . 2 H2 O
Quantity 5 g
Synonym Ethanedioic Acid Potassium Salt Hydrate, Potassium Oxalate Dihydrate, Potassium Oxalate Dihydrate, Potassium Tetroxalate Dihydrate, Oxalic Acid Hemipotassium Salt
InChI Formula InChI=1S/2C2H2O4.K.2H2O/c2*3-1(4)2(5)6;;;/h2*(H,3,4)(H,5,6);;2*1H2/q;;+1;;/p-1
SMILES O.O.[K+].OC(=O)C(=O)O.OC(=O)C(=O)[O-]
IUPAC Name potassium;2-hydroxy-2-oxoacetate;oxalic acid;dihydrate
Molecular Weight (g/mol) 254.1907
Formula Weight 253.9676
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.