missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid, Mikromol™

Product Code. 17900836
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17900836 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17900836 Supplier Mikromol Supplier No. MM0928.01
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Oxalic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxalic Acid
CAS 144-62-7
Analyte or Component Names Oxalic Acid
Shipping Condition Room Temperature
Molecular Formula C2 H2 O4
Certifications/Compliance ISO 9001
Synonym Oxalic Acid, Ethanedioic acid, Oxalic acid, Aktisal, Aquisal, DeerClean, NSC 132055, NSC 151956, NSC 62774, NSC 76990, Ultraplast Activate S 52, Oxaliplatin Related Compound A, Ascorbic Acid Impurity E, Sodium Ascorbate Impurity E
InChI Formula InChI=1S/C2H2O4/c3-1(4)2(5)6/h(H,3,4)(H,5,6)
SMILES OC(=O)C(=O)O
IUPAC Name oxalic acid
Molecular Weight (g/mol) 90.03
Quantity 500 mg
Formula Weight 89.9953 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.