missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid, LoGiCal

Product Code. 30460268
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460268 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460268 Supplier LoGiCal Supplier No. LGCFOR0928.01
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxalic Acid
CAS 144-62-7
Analyte or Component Names Oxalic Acid
Shipping Condition Room Temperature
Molecular Formula C2 H2 O4
Certifications/Compliance ISO 9001
Synonym Oxalic Acid, Ethanedioic acid, Oxalic acid, Aktisal, Aquisal, DeerClean, NSC 132055, NSC 151956, NSC 62774, NSC 76990, Ultraplast Activate S 52, Oxaliplatin Related Compound A, Ascorbic Acid Impurity E, Sodium Ascorbate Impurity E
InChI Formula InChI=1S/C2H2O4/c3-1(4)2(5)6/h(H,3,4)(H,5,6)
Solution Type Neat
SMILES OC(=O)C(=O)O
IUPAC Name oxalic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 90.03
Quantity 10 mg
Formula Weight 89.9953 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.