missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid dihydrate, Solstice

Product Code. 16685823
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
25 kg
Unit Size:
1kg
25kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16685823 25 kg 25kg
16695823 500 g 500g
15985920 1 kg 1kg
16605833 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16685823 Supplier Honeywell Fluka Supplier No. 33506H25KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (manganometric)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6153-56-6
PubChem CID 61373

Specifications

Percent Purity 99.5% (min.)
Grade ACS
Quantity 25 kg
Loss on Drying 28 - 30 %
Packaging Poly Drum
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.