missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid Dihydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10111820 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 10111820

Brand: Fisher Chemical O/0650/65

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) 126.064
MDL Number 149102
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O

Specifications

Packaging Plastic keg
Vapor Pressure 21.5mbar 50
Physical Form Powder/Solid
Color White
pH 1.3
Melting Point 98°C
Quantity 5 kg
Formula Weight 126.04g/mol
compliance-icons compliance-icons
Product Identifier
  • Oxalic acid dihydrate
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.