missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid dihydrate, ACS, 99.5-102.5%

Product Code. 11397568
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
Unit Size:
1kg
250g
50g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397568 1 kg 1kg
11387568 50 g 50g
11317578 250 g 250g
11307578 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11397568 Supplier Thermo Scientific Alfa Aesar Supplier No. 033262.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6153-56-6 | C2H6O6 | 126.064 g/mol

As an analytical reagent, in dye and paint industries, in polymers, in metal treatment, in cleaning and textile finishing and as a reagent for dehydration and condensation Oxalic acid dihydrate acts as an analytical reagent, as a reducing agent for photographic films and as mordant in the dyeing process. It is used as a plating agent, a surface treating agent, a grinding agent, a precipitating agent in rare-earth mineral processing, a bleaching agent in textile and a purifying agent in the pharmaceutical industry. It plays an important role in antibiotic medication like oxytetracycline and chloramphenicol. It finds application in water treatment plants to remove calcium waste water. It is also used as a buffer in chromatographic separation as well as bleach for wood, removing black stains caused by water penetration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • As an analytical reagent, in dye and paint industries, in polymers, in metal treatment, in cleaning and textile finishing, and as a reagent for dehydration and condensation
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) 126.064
MDL Number MFCD00149102
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O

Specifications

pH 1.3
Melting Point 101°C to 105°C
Color White
Assay Percent Range 99.5-102.5%
Linear Formula HO2CCO2H·2H2O
UN Number UN3261
Beilstein 3679436
Merck Index 14,6911
Solubility Information Soluble in water,ethanol,ether and glycerol; Insoluble in chloroform.
Formula Weight 126.07 (90.04 Anhydrous)
Percent Purity 99.5 to 102.5%
Grade ACS
Density 1.65 g/mL
Quantity 1 kg
Odor Odorless
Physical Form Crystalline
Chemical Name or Material Oxalic acid dihydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Oxalic acid dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Oxalic acid dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.