missing translation for 'onlineSavingsMsg'
Learn More

Oxalate, Ion chromatography standard solution, Specpure™, C2O4-2 1000μg/mL, Thermo Scientific Chemicals

£86.90 - £166.00

Chemical Identifiers

MDL Number MFCD00012465
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11389168
View Documents Promotion Details
Thermo Scientific Alfa Aesar
042568.AE
100 mL
£86.90
100mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11326119
View Documents Promotion Details
Thermo Scientific Alfa Aesar
042568.AP
500 mL
£166.00
500mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00012465

Specifications

MFCD00012465
Odorless
Oxalate, Ion chromatography standard solution, C2O4-2 1000μg/mL
Specpure™
Liquid
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only