Learn More
Oxalacetic acid, 98+%, Thermo Scientific Chemicals
£32.70 - £112.00
Chemical Identifiers
CAS | 328-42-7 |
---|---|
Molecular Formula | C4H4O5 |
Molecular Weight (g/mol) | 132.071 |
MDL Number | MFCD00002592 |
InChI Key | KHPXUQMNIQBQEV-UHFFFAOYSA-N |
Synonym | oxalacetic acid, oxaloacetic acid, 2-oxosuccinic acid, oxaloacetate, ketosuccinic acid, oxosuccinic acid, 2-ketosuccinic acid, butanedioic acid, oxo, oxobutanedioic acid, oxalacetate |
PubChem CID | 970 |
ChEBI | CHEBI:30744 |
IUPAC Name | 2-oxobutanedioic acid |
SMILES | C(C(=O)C(=O)O)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11367138
|
Thermo Scientific Alfa Aesar
015789.04 |
2 g |
£32.70
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11377138
|
Thermo Scientific Alfa Aesar
015789.09 |
10 g |
£112.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Oxaloacetic acid is a substrate for malate dehydrogenase and oxaloacetate decarboxylase. It is an inhibitor of succinic dehydrogenase. It is an intermediate in the citric acid cycle and glucogenesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
328-42-7 | |
132.071 | |
KHPXUQMNIQBQEV-UHFFFAOYSA-N | |
970 | |
2-oxobutanedioic acid |
C4H4O5 | |
MFCD00002592 | |
oxalacetic acid, oxaloacetic acid, 2-oxosuccinic acid, oxaloacetate, ketosuccinic acid, oxosuccinic acid, 2-ketosuccinic acid, butanedioic acid, oxo, oxobutanedioic acid, oxalacetate | |
CHEBI:30744 | |
C(C(=O)C(=O)O)C(=O)O |
Specifications
328-42-7 | |
88°C (190°F) | |
C4H4O5 | |
UN3261 | |
14,6909 | |
Soluble in water (0.1g/ml),ether,ethanol,acetone,and acetone ethyl acetate. | |
C(C(=O)C(=O)O)C(=O)O | |
132.071 | |
CHEBI:30744 | |
≥98% | |
Oxalacetic acid |
161°C (decomposition) | |
>98% | |
MFCD00002592 | |
1705475 | |
oxalacetic acid, oxaloacetic acid, 2-oxosuccinic acid, oxaloacetate, ketosuccinic acid, oxosuccinic acid, 2-ketosuccinic acid, butanedioic acid, oxo, oxobutanedioic acid, oxalacetate | |
KHPXUQMNIQBQEV-UHFFFAOYSA-N | |
2-oxobutanedioic acid | |
970 | |
132.07 | |
Powder |
Safety and Handling
GHS H Statement
H314-H318
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 206-329-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only