missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalacetic acid, 98+%, Thermo Scientific Chemicals
74.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
2 g
10 g
Description
Oxaloacetic acid is a substrate for malate dehydrogenase and oxaloacetate decarboxylase. It is an inhibitor of succinic dehydrogenase. It is an intermediate in the citric acid cycle and glucogenesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 161°C (decomposition) |
Flash Point | 88°C (190°F) |
Assay Percent Range | >98% |
Quantity | 10 g |
UN Number | UN3261 |
Beilstein | 1705475 |
Merck Index | 14,6909 |
Solubility Information | Soluble in water (0.1g/ml),ether,ethanol,acetone,and acetone ethyl acetate. |
Formula Weight | 132.07 |
Percent Purity | ≥98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'