missing translation for 'onlineSavingsMsg'
Learn More

Oxalacetic acid, 97%

Product Code. 11490693
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490693 25 g 25g
11480693 5 g 5g
11400703 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490693 Supplier Thermo Scientific Alfa Aesar Supplier No. A12739.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 328-42-7 | C4H4O5 | 132.071 g/mol

Oxalacetic acid is used as a metabolic intermediate in many processes that occur in animals and citric acid cycle. It is a used to converted to aspartic acid by aspartate transaminase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxalacetic acid is used as a metabolic intermediate in many processes that occur in animals and citric acid cycle. It is a used to converted to aspartic acid by aspartate transaminase.

Solubility
Soluble in water (0.1 g/ml), ether, ethanol, acetone, and acetone ethyl acetate.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from heat and sunlight.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328-42-7
Molecular Formula C4H4O5
Molecular Weight (g/mol) 132.071
MDL Number MFCD00002592
InChI Key KHPXUQMNIQBQEV-UHFFFAOYSA-N
Synonym oxalacetic acid, oxaloacetic acid, 2-oxosuccinic acid, oxaloacetate, ketosuccinic acid, oxosuccinic acid, 2-ketosuccinic acid, butanedioic acid, oxo, oxobutanedioic acid, oxalacetate
PubChem CID 970
ChEBI CHEBI:30744
IUPAC Name 2-oxobutanedioic acid
SMILES C(C(=O)C(=O)O)C(=O)O

Specifications

Melting Point 161°C (decomposition)
Flash Point 88°C (190°F)
Quantity 25 g
UN Number UN3261
Beilstein 1705475
Merck Index 14,6909
Solubility Information Soluble in water (0.1g/ml),ether,ethanol,acetone,and acetone ethyl acetate.
Formula Weight 132.07
Percent Purity 97%
Chemical Name or Material Oxalacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.