missing translation for 'onlineSavingsMsg'
Learn More

Oxacillin sodium salt, MedChemExpress

Product Code. 16458516
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16458516 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458516 Supplier Medchem Express Supplier No. HYB0925

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Oxacillin sodium salt is a narrow-spectrum β-lactam antibiotic of the penicillin class.

Oxacillin sodium salt is a narrow-spectrum β-lactam antibiotic of the penicillin class.

Specifications

Chemical Name or Material Oxacillin sodium salt
Physical Form Solid
CAS 1173-88-2
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H18N3NaO5S
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 125 mg/mL (295.22 mM; Need ultrasonic)
SMILES O=C(O[Na])[C@@H](C(C)(C)S[C@]1([H])[C@@H]2NC(C3=C(C)ON=C3C4=CC=CC=C4)=O)N1C2=O
Molecular Weight (g/mol) 423.42
Formula Weight 423.42
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.