missing translation for 'onlineSavingsMsg'
Learn More

OXA1L Mouse anti-Human, Clone: 4A2D9, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen OXA1L
Clone 4A2D9
Concentration 0.82 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16837725
View Documents
Proteintech
66128-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16827725
View Documents
Proteintech
66128-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

OXA1L is required for the insertion of integral membrane proteins into the mitochondrial inner membrane. It is essential for the activity and assembly of cytochrome oxidase and is required for the correct biogenesis of ATP synthase and complex I in mitochondria.
Specifications

Specifications

OXA1L
0.82 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Hsa, OXA1 like protein, OXA1Hs, OXA1L
OXA1L
IgG1
Protein G
Antibody
4A2D9
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human
Q15070
5018
OXA1L Fusion Protein Ag13819
Primary
-20°C
OXA1L
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.