missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OXA1L Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93729-0.1ml
This item is not returnable.
View return policy
Description
OXA1L Polyclonal antibody specifically detects OXA1L in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| OXA1L | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| hsa, MGC133129, mitochondrial inner membrane protein OXA1L, OXA1, OXA1Hs, OXA1-like protein, oxidase (cytochrome c) assembly 1-like, Oxidase assembly 1-like protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 336-435 of human OXA1L (NP_005006.4). PAVRTVLKIPQRVVHDLDKLPPREGFLESFKKGWKNAEMTRQLREREQRMRNQLELAARGPLRQTFTHNPLLQPGKDNPPNIPSSSSKPKSKYPWHDTLG | |
| 0.1 mL | |
| Stem Cell Markers | |
| 5018 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction