missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OX40 Recombinant Rabbit Monoclonal Antibody (055), FITC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£36.72 - £113.00
Specifications
| Antigen | OX40 |
|---|---|
| Clone | 055 |
| Concentration | 100 μg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30019968
|
Invitrogen™
MA552428 |
25 Tests |
£36.72
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30019873
![]() |
Invitrogen™
MA552427 |
100 Tests |
£113.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OX40 Recombinant Monoclonal antibody specifically detects OX40 in Mouse samples. It is validated for Flow CytometrySpecifications
| OX40 | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.05% BSA and 0.03% ProClin 300 | |
| ACT35; ACT35 antigen; ATC35 antigen; CD134; CD134 antigen; IMD16; Ly-70; lymphoid activation antigene ACT35; MRC OX40; Ox40; OX40 antigen; OX40 cell surface antigen; OX40 homologue; OX40L receptor; TAX transcriptionally-activated glycoprotein 1 receptor; tax-transcriptionally activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 receptor; TNF receptor superfamily member 4; Tnfrsf4; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor receptor superfamily member 4; tumor necrosis factor receptor superfamily, member 4; tumor necrosis factor superfamily, member 4; Txgp1; Txgp1l | |
| Recombinant Mouse OX40 Protein (Val 20-Pro211). | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Tnfrsf4 |
| 055 | |
| Flow Cytometry | |
| FITC | |
| Rabbit | |
| Mouse | |
| P47741 | |
| 22163 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title