missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OX40 Monoclonal Antibody (OX40), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517498
This item is not returnable.
View return policy
Description
MA5-17498 targets CD134/OX-40 in FACS and IHC (F) applications and shows reactivity with Rat samples. The MA5-17498 immunogen is phytohaemagglutinin-activated rat lymph node cells.
OX40 is a protein receptor found on the surface of T cells, a type of white blood cell that plays a crucial role in the immune system. When activated, OX40 promotes the proliferation and survival of T cells, leading to a stronger immune response against cancer cells and infectious agents. OX40 agonists, which are drugs that activate OX40, have shown promising results in preclinical and clinical studies as a potential immunotherapy for cancer. They can enhance the efficacy of other cancer treatments such as chemotherapy and checkpoint inhibitors, and may also have applications in autoimmune diseases and allergies.
Specifications
| OX40 | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
| P15725 | |
| Tnfrsf4 | |
| Phytohaemagglutinin-activated rat lymph node cells. | |
| 100 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Frozen) | |
| OX40 | |
| FITC | |
| Tnfrsf4 | |
| ACT35; ACT35 antigen; ATC35 antigen; CD134; CD134 antigen; IMD16; Ly-70; lymphoid activation antigene ACT35; MRC OX40; Ox40; OX40 antigen; OX40 cell surface antigen; OX40 homologue; OX40L receptor; TAX transcriptionally-activated glycoprotein 1 receptor; tax-transcriptionally activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 receptor; TNF receptor superfamily member 4; Tnfrsf4; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor receptor superfamily member 4; tumor necrosis factor receptor superfamily, member 4; tumor necrosis factor superfamily, member 4; Txgp1; Txgp1l | |
| Mouse | |
| Protein G | |
| RUO | |
| 25572 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction