missing translation for 'onlineSavingsMsg'
Learn More

OX40 Ligand/TNFSF4 Antibody (R4930 (Oxelumab)) - Azide and BSA Free, Novus Biologicals™

Product Code. 18352590 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18352590 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18352590 Supplier Novus Biologicals Supplier No. NBP275913SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

OX40 Ligand/TNFSF4 Monoclonal specifically detects OX40 Ligand/TNFSF4 in Human samples. It is validated for Flow Cytometry, Block/Neutralize.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OX40 Ligand/TNFSF4
Applications Flow Cytometry, Blocking Assay, Neutralization
Classification Monoclonal
Clone R4930 (Oxelumab)
Conjugate Unconjugated
Dilution Flow Cytometry, Block/Neutralize
Formulation PBS with 0.02% Proclin 300
Gene Alias biosimilar, biosimilars, CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), ta
Gene Symbols TNFSF4
Host Species Human
Immunogen Oxelumab binds to human OX40L, the ligand of OX40, stimulatory receptor that is expressed on activated T cells, natural killer (NK) cells and natural killer T (NKT) cells.
Purification Method Protein A purified
Quantity 0.025 mg
Primary or Secondary Primary
Gene ID (Entrez) 7292.0
Test Specificity Binds to human OX40L.
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.