missing translation for 'onlineSavingsMsg'
Learn More

OX40 Ligand/TNFSF4 Antibody (R4930 (Oxelumab)), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18325520 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18325520 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18325520 Supplier Novus Biologicals Supplier No. NBP275913AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

OX40 Ligand/TNFSF4 Monoclonal specifically detects OX40 Ligand/TNFSF4 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OX40 Ligand/TNFSF4
Applications Flow Cytometry
Classification Monoclonal
Clone R4930 (Oxelumab)
Conjugate Alexa Fluor 594
Dilution Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1
Gene Symbols TNFSF4
Host Species Human
Immunogen Oxelumab binds to human OX40L, the ligand of OX40, stimulatory receptor that is expressed on activated T cells, natural killer (NK) cells and natural killer T (NKT) cells.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Cell Cycle and Replication, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 7292.0
Test Specificity Binds to human OX40L.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.