missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OX40 Ligand/TNFSF4 Antibody (oxelumab), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28571CL1
This item is not returnable.
View return policy
Description
OX40 Ligand/TNFSF4 Monoclonal antibody specifically detects OX40 Ligand/TNFSF4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| OX40 Ligand/TNFSF4 | |
| Monoclonal | |
| CoraFluor 1 | |
| CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1 | |
| TNFSF4/ OX40L/ CD252 | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Immunology | |
| 7292 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| oxelumab | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction