missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OX40 Ligand/TNFSF4 Antibody (AP-MAB0834) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-06683
This item is not returnable.
View return policy
Description
OX40 Ligand/TNFSF4 Monoclonal specifically detects OX40 Ligand/TNFSF4 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications
| OX40 Ligand/TNFSF4 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready, Block/Neutralize | |
| CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1 | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| CD252 (AP-MAB0834) | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2 |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), CyTOF, Neutralization | |
| AP-MAB0834 | |
| Unconjugated | |
| PBS with No Preservative | |
| TNFSF4 | |
| Recombinant mouse OX-40 ligand | |
| 0.5 mg | |
| Cell Cycle and Replication, Immunology | |
| 7292 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction