missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OX1R/OX2R Polyclonal Antibody
GREENER_CHOICE

Product Code. 11306933
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11306933 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11306933 Supplier Invitrogen™ Supplier No. PA518952

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein selectively binds the hypothalamic neuropeptide orexin A. A related gene encodes a G-protein coupled receptor that binds orexin A and orexin B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OX1R/OX2R
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Hcrtr2
Gene Accession No. O43613, O43614, P56718, P56719, P58307, P58308
Gene Alias Hcrtr1; Hcrtr2; Hctr1; hypocretin (orexin) receptor 1; hypocretin (orexin) receptor 2; hypocretin receptor 1; hypocretin receptor 2; hypocretin receptor type 1; Hypocretin receptor type 2; hypocretin receptor-1; mOX2aR; mOX2bR; Mox2r; mOXR2; orexin receptor 1; orexin receptor 2; orexin receptor type 1; Orexin receptor type 2; orexin receptor-1; orexin receptor-2; orexin type-2 receptor; Ox1r; ox-1-R; ox1-R; ox2r; Ox-2-R; ox2-R
Gene Symbols HCRTR1, Hcrtr2
Host Species Goat
Immunogen Synthetic peptide sequence (YNFLSGKFREQFK) corresponding to the internal amino acids of HCRTR1 (aa 358-370).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230777, 25593, 25605, 3061, 3062, 387285
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.