missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ P7-CST Multiple Gradient Gel Caster

Cast one to five gels simultaneously with the Thermo Scientific™ Owl™ P7-CST Multiple Gradient Gel Caster.

Brand:  Thermo Scientific™ P7-CST

Code : 04

Additional Details : Weight : 8.82000kg

 View more versions of this product

Product Code. 12414697

  • £204.00 / Each
Estimated Shipment: 23-07-2024
to see stock.
Add to basket

Description

Description

Cast one to five gels simultaneously with the Thermo Scientific™ Owl™ P7-CST Multiple Gradient Gel Caster. One of the simplest devices to cast gradient gels, this caster is compatible with the Owl™ PBDS system.

  • With an external gradient maker, gels can be cast with ease and great reproducibility
  • The design and format of this unit will provide years of service in your lab
Specifications

Specifications

P7-CST Gradient Gel Caster
Owl
13 cm
13 cm
Owl P7-CST System
Caster
15 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers