Learn More
Thermo Scientific™ Owl™ EasyCast™ B1A Mini Gel and A2-OK Multi Gel Electrophoresis System Combs
Product Code. 11992945
Click to view available options
Type:
Double-sided Comb
Prep Comb
Teeth Count:
2
6
8
10
12
Thickness (Metric) Teeth:
1.0/1.5 mm
1.5 mm
Unit Size:
Each
Product Code. 11992945
Brand: Thermo Scientific™ B1A12
Only null left
Add to basket
Add to basket
Eliminate comb assembly with this assortment of heavy-duty, one-piece combs.
Eliminate comb assembly with Thermo Scientific™ Owl™ EasyCast™ B1A Mini Gel and A2-OK Multi Gel Electrophoresis System Combs. This assortment of heavy-duty, one-piece combs is for use with the EasyCast B1A Mini Gel and A2-OK Multi Gel Electrophoresis System.
Ordering Information:
Customized combs are available (order SKU#: XCM). Please contact your customer service or sales representative for further details.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl™ EasyCast B1A Mini Gel and A2-OK Multi Gel Electrophoresis System
Gel Size Mini
Product Line Owl, EasyCast
Type Double-sided Comb
Teeth Count 12
Thickness (Metric) Teeth 1.0/1.5 mm