Learn More
Thermo Scientific™ Owl™ EasyCast™ B1A Mini Gel and A2-OK Multi Gel Electrophoresis System Combs
Eliminate comb assembly with this assortment of heavy-duty, one-piece combs.
Brand: Thermo Scientific™ B1A-6
Code : 04
Additional Details : Weight : 0.00800kg
Description
Eliminate comb assembly with Thermo Scientific™ Owl™ EasyCast™ B1A Mini Gel and A2-OK Multi Gel Electrophoresis System Combs. This assortment of heavy-duty, one-piece combs is for use with the EasyCast B1A Mini Gel and A2-OK Multi Gel Electrophoresis System.
Ordering Information:
Customized combs are available (order SKU#: XCM). Please contact your customer service or sales representative for further details.
Specifications
6 tooth comb, 1.0/1.5 mm Double Sided | |
Mini | |
Comb | |
1.0/1.5 mm |
Owl™ EasyCast B1A Mini Gel and A2-OK Multi Gel Electrophoresis System | |
Owl, EasyCast | |
6 |