missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System
missing translation for 'orderingAttributeHoverText'
Includes:
Without Buffer Exchange Ports
missing translation for 'unitSize'
Each
Descripción
Obtain flat, even banding patterns while conserving valuable bench space with the Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System.
Features:
- Designed to provide flat, even banding patterns and consistent results with no tape, grease, agarose seals or other accessories.
- Can run one gel tray (51 samples) in the buffer chamber, or to save bench space, two stacked trays (102 samples).
- Two 16 x 17cm UV Transmissible (UVT) gel trays accommodate up to three combs each, allowing the user to run up to three series of samples equal distances.
- Stand-alone casting platform included for casting two gels simultaneously.
- A single gel can be cast right in the buffer chamber or in the external caster.
Includes:
- Buffer chamber (buffer volume, 1.8L)
- Three-point leveling base
- Thermo Scientific™ SuperSafe™ lid with attached power supply leads
- Two UVT gel trays
- Six 1.5mm thick combs
- External gel caster
- Bubble level
Especificaciones
Especificaciones
| Certifications/Compliance | CE marked |
| Product Line | Owl |
| Type | Horizontal Electrophoresis System |
| Includes | Without Buffer Exchange Ports |
| Sample Size | 17 to 102 |
| Volume (Metric) Buffer | 1.8 L |
| Dimensions (L x W x H) | 9.1 x 10.6 x 5.5 in. (23 x 27 x 14 cm) |
| Length (English) | 9.1 in. |
| Length (Metric) | 23 cm |
| Length (Metric) Gel | 17 cm |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
