missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Replacement Parts
Description
Replace your UVT Gel Tray, gaskets and external caster with Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Replacement Parts, for use with the Owl D4 System.
- System design allows you to run one gel tray (51 samples) or stack two gel trays (102 samples) in the buffer chamber while saving valuable bench space.
- Each of the (2) U.V. Transmissible (UVT) gel trays, 16 x 17 cm (W x L), accommodates up to three combs, allowing the user to run up to three series of samples of equal distances.
Specifications
Specifications
| For Use With (Equipment) | Owl™ D4 Electrophoresis System |
| Product Line | Owl |
| Type | Gel Tray |
| Length (Metric) | 17 cm |
| Width (Metric) | 16 cm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?