Learn More
Thermo Scientific™ Owl™ D3-14 System Replacement Parts
Product Code. 11977515
Click to view available options
Type:
Caster
Gasket
Gel Tray
Unit Size:
Each
Pack of 2
Product Code. 11977515
Brand: Thermo Scientific™ D3GK
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Choose from various Thermo Scientific™ Owl™ D3-14 System Replacement Parts including UVT gel tray, gaskets and external caster.
Choose from various Thermo Scientific™ Owl™ D3-14 System Replacement Parts, including UVT gel tray, gaskets and external caster. For use with the Owl D3-14 System.
  • UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
  • Comb slots are positioned to enable up to 12 sample sets on the same gel
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl™ D3 Series Horizontal Electrophoresis System
Product Line Owl
Type Gasket
Quantity 1 Pair