Learn More
Thermo Scientific™ Owl™ D3-14 System Replacement Parts
Product Code. 11912975
Click to view available options
Type:
Caster
Gasket
Gel Tray
Unit Size:
Each
Pack of 2
Product Code. 11912975
Brand: Thermo Scientific™ D3CST14
Only null left
Add to basket
Add to basket
Choose from various Thermo Scientific™ Owl™ D3-14 System Replacement Parts including UVT gel tray, gaskets and external caster.
Choose from various Thermo Scientific™ Owl™ D3-14 System Replacement Parts, including UVT gel tray, gaskets and external caster. For use with the Owl D3-14 System.
  • UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
  • Comb slots are positioned to enable up to 12 sample sets on the same gel
TRUSTED_SUSTAINABILITY
Specifications
Description External caster (no trays/combs)
For Use With (Equipment) Owl™ D3 Series Horizontal Electrophoresis System
Product Line Owl
Type Caster