Learn More
Thermo Scientific™ D3-14, A3-1 and A6 Wide Gel Electrophoresis System Wall Comb, 1 Tooth
Product Code. 11997585
Click to view available options
Teeth Count:
25 (1X)
50 (2X)
Thickness (Metric) Teeth:
1 mm
1.5 mm
Unit Size:
Each
Product Code. 11997585
Brand: Thermo Scientific™ D3MT2C
Only null left
Add to basket
Add to basket
Eliminate comb assembly with Fisherbrand™ D3-14, A3-1 and A6 Wide-Gel Electrophoresis System Combs.
Eliminate comb assembly with Fisherbrand™ D3-14, A3-1 and A6 Wide-Gel Electrophoresis System Combs. These microwell and wall combs are heavy-duty, one-piece units for use with the D3-14, A3-1 and A6 wide gel electrophoresis systems.
Heavy-duty, one-piece combs eliminate comb assembly.
Ordering Information:
Cat. Nos. D3-MTC, -MTD, -MT2C and -MT2D are 8 and 12-channel pipette format (1X) and (2X) combs
Customized combs are available (Order SKU # XCM). Please contact your Customer Service or Sales Representative for further details.
TRUSTED_SUSTAINABILITY
Specifications
Description Microwell comb
For Use With (Equipment) Owl™ D3-14, A3-1 and A6 Wide Gel Electrophoresis System
Type Comb
Teeth Count 50 (2X)
Thickness (Metric) Teeth 1 mm