Learn More
Thermo Scientific™ Owl™ D2 Wide-Gel System Replacement Parts
Product Code. 12414607 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Caster
Gasket
Gel Tray
Unit Size:
Each
Product Code. Type unitSize
12414607 Caster Each
12424607 Gasket Each
12464607 Gel Tray Each
3 options
Product Code. 12414607
Brand: Thermo Scientific™ D2CST
Only null left
Add to basket
Add to basket
Replace the UVT gel tray, gaskets and external caster with Thermo Scientific™ Owl™ Wide-Gel System Replacement Parts, for use with Owl™ D2 Wide-Gel System.
Replace the UVT gel tray, gaskets and external caster with Thermo Scientific™ Owl™ D2 Wide-Gel System Replacement Parts, for use with Owl™ D2 Wide-Gel System.
  • UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
  • Comb slots are positioned so you can arrange up to 12 sample sets on the same gel
TRUSTED_SUSTAINABILITY
Specifications
Description External caster (no trays/combs)
For Use With (Equipment) Owl™ D2 Wide Gel Electrophoresis System
Product Line Owl
Type Caster