Learn More
Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Replacement Parts
Replace casting slides, casting tray or buffer chamber with Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Replacement Parts.
Brand: Thermo Scientific™ C2-CST
Code : 04
Additional Details : Weight : 2.21000kg
Description
Replace casting slides, casting tray or buffer chamber with Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Replacement Parts. Parts are for use with the Owl C2-S Micro Electrophoresis System, a system designed for quick restriction fragment analysis.
- Glass slides, 5 x 7.5 cm, are used as the gel tray
- Thin gels can be poured and formed by surface tension directly on the glass slide
Specifications
Gel casting tray | |
Owl™ C2-S Micro Electrophoresis System | |
Owl™ | |
Caster |