missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ A6 Wide Gel Horizontal Electrophoresis System

Product Code. 11977585
Click to view available options
Gel Size:
23 x 25 cm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11977585

Brand: Thermo Scientific™ A6

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Produce clear, tight banding patterns with the Thermo Scientific™ Owl™ A6 Wide Gel Horizontal Electrophoresis System, an ideal system for high-throughput laboratories.

Features:
  • Screens 25 to 500 samples on a single agarose gel, producing clear, tight banding patterns with no “smiling.”
  • Unique comb-slot design of the gel tray allows use of up to 10 evenly spaced combs.
  • Using special combs and a multichannel pipetter, samples may be loaded directly from a 96, eight or 12-well plate.
  • Incorporates Owl's end-gate casting design—gel tray is fitted with gasketed end gates which provide a leakproof seal without tape.
  • Sample loading with multichannel pipetter speeds up transfer.
  • Bubble level ensures casting of flat, uniformly thick gels.

Includes:

  • Buffer chamber with three-point leveling base
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • UVT gel tray with gasketed end gates
  • Four combs (two 25-well and two 50-well, 1.5mm thick)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE marked
Description Gel size: 23 x 25cm; Buffer volume: 3L; L x W x H: 35 x 38 x 11cm
Gel Size 23 x 25 cm
Product Line Owl
Type Wide
Model Model A6
Sample Size 25 to 500
Volume (Metric) Buffer 3 L
Dimensions (L x W x H) 13.75 x 14.94 x 4.38 in. (35 x 38 x 11 cm)
Length (English) 13.75 in.
Length (Metric) 35 cm
Length (Metric) Gel 25 cm
Width (English) 14.94 in.
Width (Metric) 38 cm
Width (Metric) Gel 23 cm
Height (English) 4.38 in.
Height (Metric) 11 cm
Max. Voltage 250 V
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.