Learn More
Owl™ A2-OK Multiple Gel System Gel Tray Holder
Click to view available options
For Use With (Equipment):
Owl A2 and A5 Large Gel Electrophoresis System
Unit Size:
Each
Description
Increase the ease of using multiple gels with the Thermo Scientific™ Owl™ A2-OK Multiple Gel System Gel Tray Holder, for use with the Owl A2-OK Multiple Gel System.
- Run up to six mini gels (7L x 8cmW) for photodocumentation
Specifications
Specifications
| Description | Gel tray holder for A2-OK large gel system |
| For Use With (Equipment) | Owl A2 and A5 Large Gel Electrophoresis System |
| Gel Size | Mini |
| Product Line | Owl |
| Type | Gel Tray Holder |