missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ A2 Large Gel Systems
Handle high throughput with consistent, reliable results with Thermo Scientific™ Owl™ A2 Large Gel Systems, ideal for detailed RNA/DNA analysis.
Brand: Thermo Scientific™ A2
Description
Handle high throughput with consistent, reliable results with Thermo Scientific™Owl™ A2 Large Gel Systems, ideal for detailed RNA/DNA analysis.
Features:
- Gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run
- Optional built-in buffer exchange ports allow for buffer recirculation on extended runs
- UVT gel tray has gasketed end gates for leakproof seal without tape
- Built-in leveling ensures flat, uniformly thick gels
- Comb slots allow two, three or four sample sets of equal lengths to be run on the same gel
- Microwell-format combs provide fast, error-free sample loading with a multichannel pipetter
- Samples may be loaded directly from a 96-well plate, 8 or 12 at a time
Includes:
- Buffer chamber with three-point leveling base
- Thermo Scientific™ SuperSafe™ lid with attached power supply leads
- UVT gel tray
- Gasketed end gates
- Three combs (16-, 24- and 36-well), 1.5mm thick
Specifications
CE marked | |
Owl | |
8 to 432 (16-, 24- and 36-well combs) | |
14.8 x 12.8 x 4.1 in. (37.5 x 32.5 x 10.5 cm) | |
37.5 cm | |
12.8 in. | |
20 cm | |
10.5 cm |
Complete system | |
Large Gel Electrophoresis System | |
2.3 L | |
14.8 in. | |
25 cm | |
32.5 cm | |
4.1 in. |